Tisk…

Chlorid stříbrný, ultra dry ≥99.997% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování


Ampuled under argon

35715.03 35715.06
35715.03EA 6220 CZK
35715.03 35715.06
Chlorid stříbrný, ultra dry ≥99.997% (metals basis)
Chlorid stříbrný

Purity: 99.997% CAS No.: 7783-90-6 Molecular Formula: AgCl Formula Weight: 143.32 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 5.56 Refractive Index: 2.071 Sensitivity: Light Sensitive Appearance: White Odour: Odourless Melting Point: 455° Boiling Point: 1550° Merck Reference: 14,8509 UN No.: 3077 MDL No.: MFCD00003399

Vzorec: AgCl
Bod varu: 1550 °C
Bod tání: 455 °C
Hustota: 5,56
Storage Temperature: Ambient
MDL Number: MFCD00003399
CAS číslo: 7783-90-6
EINECS: 232-033-3
UN: 3077
ADR: 9,III
Merck Index: 14,8509

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR