Dipraseodymium trioxide ≥99.9% (metals basis)
Dodavatel: Thermo Scientific
|
Varování
|
35663.09
35663.18
35663.09EA
4330
CZK
35663.09
35663.18
Dipraseodymium trioxide ≥99.9% (metals basis)
Dipraseodymium trioxide
Purity: 99.9%
CAS No.: 12036-32-7
Molecular Formula: Pr2O3
Formula Weight: 329.83
Physical Form: 3-12mm Sintered Lump
Density: 7.07
Melting Point: 2300°
Merck Reference: 14,7712
MDL No.: MFCD00011178
Vzorec:
O₃Pr₂ MW: 329,81 g/mol Bod tání: 2300 °C Hustota: 7,07 Storage Temperature: Ambient |
MDL Number:
MFCD00011178 CAS číslo: 12036-32-7 EINECS: 234-857-9 Merck Index: 14,7712 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...