Tisk…

Lanthanum nitride ≥99.9%

Dodavatel: Thermo Fisher Scientific
Varování

39342.06 39342.14
39342.06EA 5980 CZK
39342.06 39342.14
Lanthanum nitride ≥99.9%
Lanthanum nitride

Purity: 99.9% CAS No.: 25764-10-7 Molecular Formula: LaN Formula Weight: 152.91 Physical Form: -60 Mesh Powder

Vzorec: LaN
MW: 152.91
Storage Temperature: Ambient
MDL Number: MFCD00049962
CAS číslo: 25764-10-7
EINECS: 247-245-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR