Tisk…

Praseodymium triiodide, ultra dry ≥99.9% (REO, rare earth oxide basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí


Ampuled under argon

39385.03 39385.06
39385.03EA 4640 CZK
39385.03 39385.06
Praseodymium triiodide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Praseodymium triiodide

Purity: 99.9+% CAS No.: 13813-23-5 Molecular Formula: I3Pr Formula Weight: 521.62 Physical Form: Crystalline/Needles, Ampouled under argon Sensitivity: Light Sensitive and Hygroscopic MDL No.: MFCD00049513

Vzorec: I₃Pr
Bod tání: 737 °C
Storage Temperature: Ambient
MDL Number: MFCD00049513
CAS číslo: 13813-23-5
EINECS: 237-466-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR