Lanthanum(III) iodide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Lanthanum triiodide
Ampuled under argon
35801.03
35801.06
35801.03EA
3540
CZK
35801.03
35801.06
Lanthanum(III) iodide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Lanthanum(III) iodide
Purity: 99.9%
CAS No.: 13813-22-4
Molecular Formula: LaI3
Formula Weight: 519.62
Physical Form: -10 Mesh Beads, Ampouled under argon
Sensitivity: Hygroscopic
MDL No.: MFCD00049470
Vzorec:
I₃La MW: 519.62 Bod tání: 772 °C Hustota: 5.6 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00049470 CAS číslo: 13813-22-4 EINECS: 237-465-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...