Tisk…

Lanthanum(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)

Dodavatel: Thermo Fisher Scientific
Varování

41414.09 41414.18
41414.09EA 2640 CZK
41414.09 41414.18
Lanthanum(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Lanthanum(III) fluoride

Purity: 99.9% CAS No.: 13709-38-1 Molecular Formula: F3La Formula Weight: 195.90 Physical Form: 3-6mm Pieces Density: 5.936 Melting Point: 1493° MDL No.: MFCD00011073


For vacuum deposition

Vzorec: F₃La
Bod varu: ∼2327 °C (1013 hPa)
Bod tání: 1493 °C
Hustota: 5,936
Storage Temperature: Ambient
MDL Number: MFCD00011073
CAS číslo: 13709-38-1
EINECS: 237-252-8
UN: 3288
ADR: 6.1,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR