Lanthanum(III) chloride, ultra dry ≥99.9% (REO, rare earth oxide basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Lanthanum chloride, Lanthanum trichloride
Ampuled under argon
35702.09
35702.18
35702.30
35702.09EA
3430
CZK
35702.09
35702.18
35702.30
Lanthanum(III) chloride, ultra dry ≥99.9% (REO, rare earth oxide basis)
Lanthanum(III) chloride
Purity: 99.9%
CAS No.: 10099-58-8
Molecular Formula: LaCl3
Formula Weight: 245.27
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 3.842
Appearance: White
Odour: Odourless
Melting Point: 860°
Boiling Point: 1000°
Merck Reference: 14,5363
UN No.: 3077
MDL No.: MFCD00011068
Vzorec:
Cl₃La Bod varu: 1000 °C Bod tání: 860 °C Hustota: 3,842 Storage Temperature: Ambient |
MDL Number:
MFCD00011068 CAS číslo: 10099-58-8 EINECS: 233-237-5 UN: 3077 ADR: 9,III Merck Index: 14,5363 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...