Tisk…

1-Isoquinolinol ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: Isocarbostyril, 1-Hydroxyisoquinoline

H29124.09 H29124.03
H29124.09EA 5910 CZK
H29124.09 H29124.03
1-Isoquinolinol ≥97%
1-Isoquinolinol

Purity: 97% CAS No.: 491-30-5 Molecular Formula: C9H7NO Formula Weight: 145.16 Melting Point: 212-215° MDL No.: MFCD00006899

Vzorec: C₉H₇NO
MW: 145,16 g/mol
Bod tání: 212…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00006899
CAS číslo: 491-30-5
EINECS: 207-732-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR