Tisk…

1,3-Indandione ≥97%

Dodavatel: Thermo Scientific

Synonyms: 1,3-Indanedione, Indan-1,3-dione, 1H-Indene-1,3(2H)-dione

A12206.06 A12206.14 A12206.22
A12206.06EA 1160 CZK
A12206.06 A12206.14 A12206.22
1,3-Indandione ≥97%
1,3-Indandione

Purity: 97% CAS No.: 606-23-5 Molecular Formula: C9H6O2 Formula Weight: 146.15 Melting Point: 128-132° Beilstein Reference: 1210061

Vzorec: C₉H₆O₂
MW: 146,15 g/mol
Bod varu: 283 °C (1013 hPa)
Bod tání: 128…132 °C
Hustota: 1,370
Storage Temperature: Ambient
MDL Number: MFCD00003779
CAS číslo: 606-23-5
EINECS: 210-109-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR