2-Indanone ≥98%
Dodavatel: Thermo Scientific
Synonyms:
beta-Hydrindone, 2-Oxoindane, 1H-Inden-2(3H)-one
A12574.06
A12574.22
A12574.06EA
1750
CZK
A12574.06
A12574.22
2-Indanone ≥98%
2-Indanone
Purity: 98%
CAS No.: 615-13-4
Molecular Formula: C9H8O
Formula Weight: 132.16
Storage Temperature: 2°C to 8°C
Flash Point: 100°(212°F)
Melting Point: 52-56°
Boiling Point: 218° dec.
Beilstein Reference: 636550
MDL No.: MFCD00003792
Vzorec:
C₉H₈O MW: 132,16 g/mol Bod varu: 218 °C (rozkl.) Bod tání: 52…56 °C Hustota: 1,0712 g/cm³ (67 °C) Bod vzplanutí: 100 °C (212 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003792 CAS číslo: 615-13-4 EINECS: 210-410-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...