Tellurium tetrachloride ≥99.9% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Tellurium(IV) chloride
12284.14
12284.22
12284.14EA
4880
CZK
12284.14
12284.22
Tellurium tetrachloride ≥99.9% (metals basis)
Tellurium tetrachloride
Purity: 99.9%
CAS No.: 10026-07-0
Molecular Formula: Cl4Te
Formula Weight: 269.41
Physical Form: Crystalline
Density: 3.01
Appearance: Yellow
Odour: Pungent
Melting Point: 224°
Boiling Point: 380°
Solubility: Decomposes in water forming TeO2 and HCl
Merck Reference: 14,9126
UN No.: 3260
MDL No.: MFCD00011262
Vzorec:
Cl₄Te MW: 269.41 Bod varu: 380 °C Bod tání: 224 Hustota: 3.01 Storage Temperature: Ambient |
MDL Number:
MFCD00011262 CAS číslo: 10026-07-0 EINECS: 233-055-6 UN: 3260 ADR: 8 II Merck Index: 14,9126 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...