Tisk…

Tellurium tetrabromide ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Tellurium(IV) bromide

75101.09 75101.18
75101.09EA 4260 CZK
75101.09 75101.18
Tellurium tetrabromide ≥99.9% (metals basis)
Tellurium tetrabromide

Purity: 99.9% CAS No.: 10031-27-3 Molecular Formula: TeBr4 Formula Weight: 447.27 Physical Form: -4 Mesh Crystalline Density: 4.31 Melting Point: 380° Boiling Point: 420° dec. Solubility: Decomposes in water forming TeO2 and HBr Merck Reference: 14,9125 UN No.: 2923 MDL No.: MFCD00049575

Vzorec: Br₄Te
Bod varu: 420 °C (rozkl.)
Bod tání: 380 °C
Hustota: 4,31
Storage Temperature: Ambient
MDL Number: MFCD00049575
CAS číslo: 10031-27-3
EINECS: 233-090-7
UN: 2923
ADR: 8,II
Merck Index: 14,9125

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR