Tisk…

Arsenic(III) telluride ≥99.999% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

41709.09 41709.18
41709.09EA 6940 CZK
41709.09 41709.18
Arsenic(III) telluride ≥99.999% (metals basis)
Arsenic(III) telluride

Purity: 99.999% CAS No.: 12044-54-1 Molecular Formula: As2Te3 Formula Weight: 532.64 Physical Form: 3-12mm Pieces Density: 6.50 Melting Point: 381° UN No.: 1557 MDL No.: MFCD00014167


Vacuum deposition

Vzorec: As₂Te₃
Bod tání: 381 °C
Hustota: 6,50
Storage Temperature: Ambient
MDL Number: MFCD00014167
CAS číslo: 12044-54-1
EINECS: 234-955-1
UN: 1557
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR