Antimony(III) telluride ≥99.999% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
36282.09
36282.18
36282.09EA
5120
CZK
36282.09
36282.18
Antimony(III) telluride ≥99.999% (metals basis)
Antimony(III) telluride
Purity: 99.999%
CAS No.: 1327-50-0
Molecular Formula: Sb2Te3
Formula Weight: 626.30
Physical Form: 3-12mm Lump
Density: 6.5
Appearance: Grey
Odour: Odourless
Melting Point: 629°
UN No.: 1549
MDL No.: MFCD00016320
For vacuum deposition
Vzorec:
Sb₂Te₃ Bod tání: 629 °C Hustota: 6,5 Storage Temperature: Ambient |
MDL Number:
MFCD00016320 CAS číslo: 1327-50-0 EINECS: 215-480-9 UN: 1549 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...