Dibismuth triselenide ≥99.999% (metals basis) for vacuum deposition
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
13126.09
13126.18
13126.09EA
3780
CZK
13126.09
13126.18
Dibismuth triselenide ≥99.999% (metals basis) for vacuum deposition
Dibismuth triselenide
Purity: 99.999%
CAS No.: 12068-69-8
Molecular Formula: Bi2Se3
Formula Weight: 654.84
Physical Form: 1-6 mm Pieces
Density: 6.82
Appearance: Grey
Melting Point: 710°
Solubility: Insoluble in water. Decomposes when heated in air. Decomposes in nitric acid and aqua regia
Merck Reference: 14,1278
UN No.: 3283
MDL No.: MFCD00014200
In semiconductor research, multilayer thin-film, magnetoresistant film
Vzorec:
Bi₂Se₃ Bod tání: 710 °C Hustota: 6,82 Storage Temperature: Ambient |
MDL Number:
MFCD00014200 CAS číslo: 12068-69-8 EINECS: 235-104-7 UN: 3283 ADR: 6.1,III Merck Index: 14,1278 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...