Diantimony triselenide ≥99.999% (metals basis), pieces
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
13130.06
13130.14
13130.06EA
6700
CZK
13130.06
13130.14
Diantimony triselenide ≥99.999% (metals basis), pieces
Diantimony triselenide
Purity: 99.999%
CAS No.: 1315-05-5
Molecular Formula: Sb2Se3
Formula Weight: 480.38
Physical Form: 3-12mm Pieces
Sensitivity: Moisture Sensitive
Solubility: Very slightly soluble in water
Merck Reference: 14,712
UN No.: 3077
MDL No.: MFCD00016319
Vzorec:
Sb₂Se₃ Bod tání: 611 °C Hustota: 5,81 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016319 CAS číslo: 1315-05-5 EINECS: 215-256-0 UN: 3077 ADR: 9,III Merck Index: 14,712 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...