Tisk…

Chlorid vizmutitý, (max. 3% H₂O) ≥98% (dry weight)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Bismuth(III) chloride, Bismuth trichloride

A10510.14 A10510.22 A10510.36
A10510.14EA 1560 CZK
A10510.14 A10510.22 A10510.36
Chlorid vizmutitý, (max. 3% H₂O) ≥98% (dry weight)
Chlorid vizmutitý

Purity: 98% CAS No.: 7787-60-2 Molecular Formula: BiCl3 Formula Weight: 315.34 Odour: Irritating Melting Point: 230-232° Boiling Point: ca 430° subl. Merck Reference: 14,1262 UN No.: 3260 MDL No.: MFCD00003461

Vzorec: BiCl₃
MW: 315.34
Bod varu: ∼430 °C (subl.)
Bod tání: 230…232 °C
Hustota: 4.75
Storage Temperature: Ambient
MDL Number: MFCD00003461
CAS číslo: 7787-60-2
EINECS: 232-123-2
UN: 3260
ADR: 8 III
Merck Index: 14,1262

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR