Tisk…

Chlorid antimonitý ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Antimony trichloride

41537.14 41537.22
41537.14EA 2170 CZK
41537.14 41537.22
Chlorid antimonitý ≥99.9% (metals basis)
Chlorid antimonitý

Purity: 99.9% CAS No.: 10025-91-9 Molecular Formula: Cl3Sb Formula Weight: 228.11 Physical Form: Crystalline Density: 3.14 Appearance: White Odour: Pungent Melting Point: 73.4° Boiling Point: 223.5° Merck Reference: 14,707 UN No.: 1733 MDL No.: MFCD00011212

Vzorec: SbCl₃
MW: 228.12
Bod varu: 223.5 °C
Bod tání: 73.4 °C
Hustota: 3.14
Storage Temperature: Ambient
MDL Number: MFCD00011212
CAS číslo: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8 II
Merck Index: 14,707

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR