Phosphoryl tribromide ≥99%
Dodavatel: abcr
|
Nebezpečí
|
S93-1586
S93-1586.0100
S93-1586.0025
ABCRS93-1586EA
0
CZK
ABCRS93-1586
ABCRS93-1586.0100
ABCRS93-1586.0025
Phosphoryl tribromide ≥99%
Phosphoryl tribromide
Vzorec:
Br₃OP Bod varu: 193 °C Bod tání: 56 °C Hustota: 2,822 Storage Temperature: Ambient |
MDL Number:
MFCD00036298 CAS číslo: 7789-59-5 EINECS: 232-177-7 UN: 1939 ADR: 8,II Merck Index: 12,07505 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...