Phosphorus(V) bromide 95%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Phosphorus pentabromide
B22481.14
B22481.22
B22481.36
B22481.14EA
2880
CZK
B22481.14
B22481.22
B22481.36
Phosphorus(V) bromide 95%
Phosphorus(V) bromide
Purity: 95%
CAS No.: 7789-69-7
Molecular Formula: PBr5
Formula Weight: 430.50
Odour: Acrid
Melting Point: ca 105° dec.
Boiling Point: 105-107° dec.
Beilstein Reference: 3903332
Merck Reference: 14,7350
UN No.: 2691
MDL No.: MFCD00011437
Vzorec:
PBr₅ Bod varu: 105…107 °C (rozkl.) Hustota: 3,61 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011437 CAS číslo: 7789-69-7 EINECS: 232-186-6 UN: 2691 ADR: 8,II Merck Index: 14,7350 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...