Bromid fosforitý ≥99% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Phosphorus tribromide
Purity: 99%
CAS No.: 7789-60-8
Molecular Formula: PBr3
Formula Weight: 270.70
Physical Form: Liquid
Density: 2.852
Refractive Index: 1.697
Sensitivity: Moisture Sensitive
Odour: Pungent
Melting Point: -40°
Boiling Point: 172.9°
Solubility: Soluble in acetone and carbon disulfide. Decomposes in water and alcohol
Beilstein Reference: 3903060
Merck Reference: 14,7357
UN No.: 1808
MDL No.: MFCD00011436
Vzorec:
PBr₃ MW: 270.69 Bod varu: 172.9 °C Bod tání: –40 °C Hustota: 2.852 Storage Temperature: Ambient |
MDL Number:
MFCD00011436 CAS číslo: 7789-60-8 EINECS: 232-178-2 UN: 1808 ADR: 8 II Merck Index: 14,7357 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...