Tisk…

Germanium(II) telluride ≥99.999%

Dodavatel: Thermo Scientific
Varování

45461.03
45461.03EA 2740 CZK
45461.03
Germanium(II) telluride ≥99.999%
Germanium(II) telluride

Purity: 99.999% CAS No.: 12025-39-7 Molecular Formula: GeTe Formula Weight: 200.19 Physical Form: -200 Mesh Powder Density: 6.14 Melting Point: 725° MDL No.: MFCD00135541

Vzorec: GeTe
MW: 200,21 g/mol
Bod tání: 725 °C
Hustota: 6,14
Storage Temperature: Ambient
MDL Number: MFCD00135541
CAS číslo: 12025-39-7
EINECS: 234-706-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR