Tisk…

Chlorid cínatý, anhydrous 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Stannous chloride, Tin dichloride

A16202.22 A16202.0E A16202.36
A16202.22EA 1340 CZK
A16202.22 A16202.0E A16202.36
Chlorid cínatý, anhydrous 98%
Chlorid cínatý

Purity: 98% CAS No.: 7772-99-8 Molecular Formula: SnCl2 Formula Weight: 189.62 Melting Point: 247° Boiling Point: 652° Merck Reference: 14,8783 UN No.: 3260 MDL No.: MFCD00011241

Vzorec: SnCl₂
Bod varu: 652 °C
Bod tání: 247 °C
Hustota: 3,95
Storage Temperature: Ambient
MDL Number: MFCD00011241
CAS číslo: 7772-99-8
EINECS: 231-868-0
UN: 3260
ADR: 8,III
Merck Index: 14,8783

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR