Tisk…

Tetrachlorsilan ≥99% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Silicon tetrachloride, Silicon (IV) chloride

69182.AB 69182.AE 69182.K2 69182.L7
69182.ABEA 893 CZK
69182.AB 69182.AE 69182.K2 69182.L7
Tetrachlorsilan ≥99% (metals basis)
Tetrachlorosilane

Purity: 99% CAS No.: 10026-04-7 Molecular Formula: Cl4Si Formula Weight: 169.90 Physical Form: Liquid Density: 1.483 Appearance: Colourless Odour: Acrid Melting Point: -70° Boiling Point: 57.6° Merck Reference: 14,8500 UN No.: 1818 MDL No.: MFCD00011229

Vzorec: Cl₄Si
Bod varu: 57,6 °C
Bod tání: –70 °C
Hustota: 1,483
Storage Temperature: Ambient
MDL Number: MFCD00011229
CAS číslo: 10026-04-7
EINECS: 233-054-0
UN: 1818
ADR: 8,II
Merck Index: 14,8500

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR