Tisk…

Gallium(II) telluride ≥99.999% (metals basis)

Dodavatel: Thermo Scientific
Varování

41881.03 41881.06
41881.03EA 5800 CZK
41881.03 41881.06
Gallium(II) telluride ≥99.999% (metals basis)
Gallium(II) telluride

Purity: 99.999% CAS No.: 12024-14-5 Molecular Formula: GaTe Formula Weight: 197.32 Physical Form: 6mm & down Sensitivity: Moisture Sensitive MDL No.: MFCD00135536

Vzorec: GaTe
MW: 197,32 g/mol
Bod varu: 103…104 °C (0,08 mmHg)
Bod tání: 824 °C
Hustota: 5,44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00135536
CAS číslo: 12024-14-5
EINECS: 234-690-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR