Kyselina boritá ≥99.5% ACS
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 99.5% min
CAS No.: 10043-35-3
Molecular Formula: BH3O3
Formula Weight: 61.83
Flash Point: None
Physical Form: Granular
Density: 1.435
Appearance: White
Odour: Odourless
pH: 3.8-4.8
Melting Point: ca 185° dec.
Solubility: Soluble in water (1 gram in 18ml cold or 4ml boiling water). Solubility in water is increased by addition of HCl, citric or tartaric acid
Beilstein Reference: 1697939
Merck Reference: 14,1336
MDL No.: MFCD00011337
In hardening steel, preservatives, manufacturing of cements, porcelain, enamels, glass, borates, artificial gems, in nickeling baths, painting, photography
Vzorec:
B(OH)₃ Bod varu: 185 °C (1013 hPa) Hustota: 1,435 Storage Temperature: Ambient |
MDL Number:
MFCD00011337 CAS číslo: 10043-35-3 EINECS: 233-139-2 REACH: 01-2119486683-25 Merck Index: 14,1336 |