Tisk…

Oxid boritý ≥97.5%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Boron oxide, Boric anhydride

12290.36 12290.A3 12290.A9
12290.36EA 1390 CZK
12290.36 12290.A3 12290.A9
Oxid boritý ≥97.5%
Oxid boritý

Purity: 97.5% min CAS No.: 1303-86-2 Molecular Formula: B2O3 Formula Weight: 69.62 Physical Form: -60 Mesh Density: 2.46 +/- 0.01 Appearance: White Odour: Odourless Melting Point: ca 450° Boiling Point: 1860° Solubility: Slightly soluble in water @ 25°C. Soluble in water @ 100°C Merck Reference: 14,1337 MDL No.: MFCD00011315


As a flux, acid catalyst, or chemical intermediate in production of boron halides, esters, carbide, nitride, and metallic borides

Vzorec: B₂O₃
MW: 69.62
Bod varu: 1860 °C
Bod tání: ∼450 °C
Hustota: 2.46
Storage Temperature: Ambient
MDL Number: MFCD00011315
CAS číslo: 1303-86-2
EINECS: 215-125-8
REACH: 01-2119486655-24
Merck Index: 13,01327

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR