Tisk…

Oxid boritý, dehydrated (max. 0.02% H₂O) ≥99.999% (metals basis), Puratronic®

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Boron oxide, Boric anhydride

11159.QE 11159.QH
11159.QEEA 3610 CZK
11159.QE 11159.QH
Oxid boritý, dehydrated (max. 0.02% H₂O) ≥99.999% (metals basis), Puratronic®
Oxid boritý

Purity: 99.999% CAS No.: 1303-86-2 Molecular Formula: B2O3 Formula Weight: 69.62 Physical Form: Cylinder, 3.7cm dia, 32g/cylinder Density: 2.46 +/- 0.01 Appearance: White Odour: Odourless Melting Point: ca 450° Boiling Point: 1860° Merck Reference: 14,1337 MDL No.: MFCD00011315


In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP)

Vzorec: B₂O₃
MW: 69.62
Bod varu: 1860 °C
Bod tání: ∼450 °C
Hustota: 2.46
Storage Temperature: Ambient
MDL Number: MFCD00011315
CAS číslo: 1303-86-2
EINECS: 215-125-8
REACH: 01-2119486655-24
Merck Index: 14,1337

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR