Thallium(I) fluoride ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
78111.14
78111.22
78111.14EA
6800
CZK
78111.14
78111.22
Thallium(I) fluoride ≥97%
Thallium(I) fluoride
Purity: 97%
CAS No.: 7789-27-7
Molecular Formula: TlF
Formula Weight: 223.37
Physical Form: Crystalline
Density: 8.36
Boiling Point: 655°
Solubility: Very soluble in water
Merck Reference: 14,9262
UN No.: 1707
MDL No.: MFCD00049607
Vzorec:
FTl MW: 223,38 g/mol Bod varu: 655 °C Bod tání: 315 °C Hustota: 8,36 Storage Temperature: Ambient |
MDL Number:
MFCD00049607 CAS číslo: 7789-27-7 EINECS: 232-154-1 UN: 1707 ADR: 6.1,II Merck Index: 14,9262 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...