Tisk…

Triethanolamine hydrochloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

T9534-100G T9534-1KG T9534-250G T9534-500G
SIALT9534-100GEA 6140 CZK
SIALT9534-100G SIALT9534-1KG SIALT9534-250G SIALT9534-500G
Triethanolamine hydrochloride, Sigma-Aldrich®
Triethanolamine hydrochloride
Vzorec: C₆H₁₆ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD00012596
CAS číslo: 637-39-8
EINECS: 211-284-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR