Triethanolamine hydrochloride, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
T1502-100G
T1502-1KG
T1502-250G
T1502-500G
SIALT1502-100GEA
2250
CZK
SIALT1502-100G
SIALT1502-1KG
SIALT1502-250G
SIALT1502-500G
Triethanolamine hydrochloride, Sigma-Aldrich®
Triethanolamine hydrochloride
Vzorec:
C₆H₁₆ClNO₃ MW: 185.65 Storage Temperature: Ambient |
MDL Number:
MFCD00012596 CAS číslo: 637-39-8 EINECS: 211-284-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...