(±)-Epoxystyrene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
S5006-100G-A
S5006-500G-A
S5006-5G-A
SIALS5006-100G-AEA
1350
CZK
SIALS5006-100G-A
SIALS5006-500G-A
SIALS5006-5G-A
(±)-Epoxystyrene, Sigma-Aldrich®
(±)-Epoxystyrene
Vzorec:
C₈H₈O |
MDL Number:
MFCD00005121 CAS číslo: 96-09-3 EINECS: 202-476-7 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...