Tisk…
3-Quinuclidinone hydrochloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: 1-Azabicyclo[2.2.2]octan-3-one hydrochloride
M-Clarity™ quality level = MQ100
Q1905-100G Q1905-25G
SIALQ1905-100GEA 11610 CZK
SIALQ1905-100G SIALQ1905-25G
3-Quinuclidinone hydrochloride, Sigma-Aldrich®
3-Quinuclidinone hydrochloride
Vzorec: C₇H₁₂ClNO
MW: 161.63
MDL Number: MFCD00137391
CAS číslo: 1193-65-3
EINECS: 214-776-5