Tisk…

3-Guanidinopropionic acid, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ200

G6878-1G G6878-5G
SIALG6878-1GEA 2020 CZK
SIALG6878-1G SIALG6878-5G
3-Guanidinopropionic acid, Sigma-Aldrich®
3-Guanidinopropionic acid
Vzorec: C₄H₉N₃O₂
MDL Number: MFCD00045939
CAS číslo: 353-09-3
EINECS: 206-530-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR