Glyoxylic acid monohydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Formylformic acid monohydrate, Oxoacetic acid monohydrate, 2-Oxoethanoic acid monohydrate
M-Clarity™ quality level = MQ200
G10601-25G
G10601-100G
G10601-10G
SIALG10601-25GEA
1450
CZK
SIALG10601-25G
SIALG10601-100G
SIALG10601-10G
Glyoxylic acid monohydrate, Sigma-Aldrich®
Glyoxylic acid monohydrate
Vzorec:
C₂H₂O₃·1H₂O MW: 92.05 |
MDL Number:
MFCD00149497 CAS číslo: 563-96-2 EINECS: 206-058-5 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...