Tisk…

Ethoxymethylenemalononitrile, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

E6200-100G E6200-25G
SIALE6200-100GEA 5370 CZK
SIALE6200-100G SIALE6200-25G
Ethoxymethylenemalononitrile, Sigma-Aldrich®
Ethoxymethylenemalononitrile
Vzorec: C₆H₆N₂O
MDL Number: MFCD00001854
CAS číslo: 123-06-8
EINECS: 204-597-0
UN: 3439
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR