Tisk…

Diallylamine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

D9603-500ML D9603-100ML D9603-1L
SIALD9603-500MLEA 2700 CZK
SIALD9603-500ML SIALD9603-100ML SIALD9603-1L
Diallylamine, Sigma-Aldrich®
Diallylamine
Vzorec: C₆H₁₁N
MDL Number: MFCD00008642
CAS číslo: 124-02-7
EINECS: 204-671-2
UN: 2359
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR