Tisk…

Diethyl chlorophosphate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

D91632-25G D91632-100G
SIALD91632-25GEA 1450 CZK
SIALD91632-25G SIALD91632-100G
Diethyl chlorophosphate, Sigma-Aldrich®
Diethyl chlorophosphate
Vzorec: C₄H₁₀ClO₃P
MDL Number: MFCD00009075
CAS číslo: 814-49-3
EINECS: 212-396-4
UN: 3278
ADR: 6.1,I

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR