Tisk…
Diethyl acetamidomalonate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Diethyl 2-acetamidomalonate
M-Clarity™ quality level = MQ100
D84601-100G D84601-25G D84601-500G
SIALD84601-100GEA 1250 CZK
SIALD84601-100G SIALD84601-25G SIALD84601-500G
Diethyl acetamidomalonate, Sigma-Aldrich®
Diethyl acetamidomalonate
Vzorec: C₉H₁₅NO₅
MW: 217.22
MDL Number: MFCD00009146
CAS číslo: 1068-90-2
EINECS: 213-952-9