Tisk…

5,5-Diphenylhydantoin, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

D4007-5G D4007-100G
SIALD4007-5GEA 1310 CZK
SIALD4007-5G SIALD4007-100G
5,5-Diphenylhydantoin, Sigma-Aldrich®
5,5-Diphenylhydantoin
Vzorec: C₁₅H₁₂N₂O₂
MW: 252,27 g/mol
MDL Number: MFCD00005264
CAS číslo: 57-41-0
EINECS: 200-328-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR