Tisk…

Diphenyl chlorophosphate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Diphenyl phosphorochloridate

M-Clarity™ quality level = MQ200

D206555-100G D206555-25G
SIALD206555-100GEA 2220 CZK
SIALD206555-100G SIALD206555-25G
Diphenyl chlorophosphate, Sigma-Aldrich®
Diphenyl chlorophosphate
Vzorec: C₁₂H₁₀ClO₃P
MW: 268,64 g/mol
MDL Number: MFCD00003030
CAS číslo: 2524-64-3
EINECS: 219-759-6
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR