Tisk…

Dimethyl acetylenedicarboxylate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Dimethyl but-2-yne-1,4-dioate, Dimethyl but-2-ynedioate

M-Clarity™ quality level = MQ200

D138401-25G D138401-5G
SIALD138401-25GEA 2330 CZK
SIALD138401-25G SIALD138401-5G
Dimethyl acetylenedicarboxylate, Sigma-Aldrich®
Dimethyl acetylenedicarboxylate
Vzorec: C₆H₆O₄
MDL Number: MFCD00008456
CAS číslo: 762-42-5
EINECS: 212-098-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR