Tisk…

3,4-Dihydro-2H-pyran, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

D106208-100ML D106208-500ML
SIALD106208-100MLEA 975 CZK
SIALD106208-100ML SIALD106208-500ML
3,4-Dihydro-2H-pyran, Sigma-Aldrich®
3,4-Dihydro-2H-pyran
Vzorec: C₅H₈O
MW: 84,12 g/mol
MDL Number: MFCD00006558
CAS číslo: 110-87-2
EINECS: 203-810-4
UN: 2376
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR