Tisk…

5-Chloroisatoic anhydride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 6-Chloro-1H-benzo[d][1,3]oxazine-2,4-dione

C48104-25G C48104-5G
SIALC48104-25GEA 0 CZK
SIALC48104-25G SIALC48104-5G
5-Chloroisatoic anhydride, Sigma-Aldrich®
5-Chloroisatoic anhydride
Vzorec: C₈H₄ClNO₃
MW: 197,58 g/mol
MDL Number: MFCD00006701
CAS číslo: 4743-17-3
EINECS: 225-255-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR