Tisk…
(±)-Allyl-2,3-epoxypropyl ether, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Allyl glycidyl ether
M-Clarity™ quality level = MQ200
A32608-100ML A32608-500ML
SIALA32608-100MLEA 1320 CZK
SIALA32608-100ML SIALA32608-500ML
(±)-Allyl-2,3-epoxypropyl ether, Sigma-Aldrich®
(±)-Allyl-2,3-epoxypropyl ether
Vzorec: C₆H₁₀O₂
MW: 114.14
MDL Number: MFCD00005143
CAS číslo: 106-92-3
EINECS: 203-442-4
UN: 2219
ADR: 3 III