Acryloyl chloride stabilised, Sigma-Aldrich®
Dodavatel: Merck
Stabilization:
Stabilised with phenothiazine
M-Clarity™ quality level = MQ200
A24109-1KG
A24109-100G
A24109-5G
SIALA24109-1KGEA
46270
CZK
SIALA24109-1KG
SIALA24109-100G
SIALA24109-5G
Acryloyl chloride stabilised, Sigma-Aldrich®
Acryloyl chloride
Vzorec:
C₃H₃ClO |
MDL Number:
MFCD00000731 CAS číslo: 814-68-6 EINECS: 212-399-0 UN: 3383 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...