Tisk…

4-Acetamidobenzaldehyde, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

A1800-25G A1800-5G
SIALA1800-25GEA 3730 CZK
SIALA1800-25G SIALA1800-5G
4-Acetamidobenzaldehyde, Sigma-Aldrich®
4-Acetamidobenzaldehyde
Vzorec: C₉H₉NO₂
MDL Number: MFCD00003380
CAS číslo: 122-85-0
EINECS: 204-579-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR