Tisk…

Acetylenedicarboxylic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

A15207-25G A15207-5G
SIALA15207-25GEA 8850 CZK
SIALA15207-25G SIALA15207-5G
Acetylenedicarboxylic acid, Sigma-Aldrich®
Acetylenedicarboxylic acid
Vzorec: C₄H₂O₄
MW: 114,06 g/mol
MDL Number: MFCD00004362
CAS číslo: 142-45-0
EINECS: 205-536-0
UN: 2928
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR