Paladium 5% on activated carbon for use as a catalyst, Sigma-Aldrich®
Dodavatel: Merck
75992-10G
75992-1G
75992-50G
SIAL75992-10GEA
3790
CZK
SIAL75992-10G
SIAL75992-1G
SIAL75992-50G
Paladium 5% on activated carbon for use as a catalyst, Sigma-Aldrich®
Palladium
Vzorec:
Pd MW: 106.420 g/mol |
MDL Number:
MFCD00011167 CAS číslo: 7440-05-3 EINECS: 231-115-6 |