Tisk…
Paladium 5% on activated carbon wetted with ca. 50% water for use as a catalyst, Sigma-Aldrich®
Dodavatel: Merck
Stabilization: Moistened with water
75991-10G 75991-50G
SIAL75991-10GEA 3580 CZK
SIAL75991-10G SIAL75991-50G
Paladium 5% on activated carbon wetted with ca. 50% water for use as a catalyst, Sigma-Aldrich®
Palladium
Vzorec: Pd
MW: 106.420 g/mol
MDL Number: MFCD00011167
CAS číslo: 7440-05-3
EINECS: 231-115-6